MilliporeSigma
HomeApplicationsWater PurificationWater for Cell & Molecular Biology
Water for Cell & Molecular Biology
Illustration of a DNA double helix with the outline highlighted in red and the cross ladder in blue, sitting on a sequencing document showing nucleotide bases represented by letters ‘ATCG’
Water purity is crucial in cell biology and molecular techniques as contaminants can significantly affect experimental outcomes and reproducibility. In cell culture, impurities can disrupt cell growth and function. In protein research, water quality may impact protein structure and affect enzymatic activity. Water impurities may also affect the quality of protein analysis or purification. Nucleases are often present in water and may degrade DNA or RNA samples and lead to false results in PCR and other genomic research techniques. 
Understanding the impact of specific water contaminants on these sensitive techniques is critical to ensure experimental reliability and consistency. Below you will find products and information to support you in selecting the optimal water quality for your sensitive biological applications.
Featured Categories
Milli-Q® IQ 7003/05/10/15 Ultrapure and Pure Water Purification System - Type 1 and Type 2 lab water purification system with production and tank units plus pure and ultrapure water dispensers.
Milli-Q® Benchtop Lab Water Purification Systems
Milli-Q® systems offer innovative water purification technologies engineered to support lab research needs, sustainability goals, and other major requirements.
Shop Products
Milli-Q® HX 7000 Central Water Purification System and SDS 500 Pure Water Tank
Milli-Q® High-Flow Lab Water Purification Systems
From a single lab to an entire building, Milli-Q® central water purification systems are connected, high-performance instruments that can reliably feed all purified water needs.
Shop Products
IPAK Gard® pretreatment pack
Milli-Q® Lab Water Purification Consumables
Find the cartridges, UV lamps, filters and polishers for your water system. Advantages of using genuine Milli-Q consumables: guarantee water quality, sustainability features (new Drain Cap, Hg-free lamps). Designed for perfect fit.
Shop Products
Connect up to 4 POD dispensers to one Milli-Q® IQ water system
Accessories for Milli-Q® Lab Water Purification Systems
Milli-Q® water system accessories support lab-specific needs, improving safety, water quality, and productivity while conserving bench space.
Shop Products
Impact of Water Quality on Cell Culture
As water is the largest component in any culture media formulation, its quality is critical for reliable and reproducible methods. Correct water quality is important when preparing cell culture media as impurities can affect media performance or cause abnormalities, such as changes in pH or color, or cause precipitates. Water should be free from substances, such as bacteria, heavy metals, endocrine disrupters, and endotoxins (pyrogens), which can effect cell growth and function. Using ultrapure water purified by ultrafiltration ensures the removal of these contaminants and supports the integrity and reproducibility of cell culture experiments.
Impact of Water Quality on Protein Research
High-purity water is crucial in protein research and proteomic studies. Water impurities can affect protein folding, stability and activity. Using optimal water purity is also essential for techniques like ELISA, Western blotting and protein purification techniques, where contaminants can compromise accuracy and lead to erroneous conclusions.
Impact of Water Quality on Genomics Research
The inherent sensitivity and specificity of many genomic techniques, such as PCR and sequencing, demands very high purity reagents.  Nucleases, even in trace quantities, can degrade DNA and RNA samples, impacting processes like nucleic acid extraction, PCR, gene editing or sequencing. Organic contaminants (e.g. humic acid, polysaccharides) may mimic the structure of nucleic acids or interact with them, while inorganic contaminants, like some metals, can interact with polymerases and reverse transcriptase, compromising the results of PCR-based techniques. These sensitive techniques require the use of nuclease-free ultrapure water to prevent degradation of DNA/RNA and to maintain results reliability and consistency.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents