MilliporeSigma
HomeApplicationsWater PurificationWater for Clinical Diagnostics
Water for Clinical Diagnostics
A gloved hand places a test tube holding a blood sample into a plastic laboratory test tube rack holding other blood samples with various colored lids
In biomedical laboratories, clinical analyzers are critical instruments that require large quantities of purified water to operate continuously in order to perform timely tests. Any water-related issue, such as a deviation in expected quality or flow rate, could cause analyzer downtime, resulting in decreased laboratory productivity and a delay in delivering results to healthcare professionals and their patients.
The information below is intended to support clinical lab managers and anyone responsible for the water purification systems in a biomedical laboratory. Read to learn how to prevent water contaminants from interfering with the delivery of accurate and reliable chemistry, hematology and immunology assays. You’ll also learn how to maintain maximum water system and analyzer uptime for more efficient clinical lab operations.
Featured Categories
Milli-Q® HX 7000 Central Water Purification System and SDS 500 Pure Water Tank
Milli-Q® High-Flow Lab Water Purification Systems
From a single lab to an entire building, Milli-Q® central water purification systems are connected, high-performance instruments that can reliably feed all purified water needs.
Shop Products
Milli-Q® IQ 7003/05/10/15 Ultrapure and Pure Water Purification System - Type 1 and Type 2 lab water purification system with production and tank units plus pure and ultrapure water dispensers.
Milli-Q® Benchtop Lab Water Purification Systems
Milli-Q® systems offer innovative water purification technologies engineered to support lab research needs, sustainability goals, and other major requirements.
Shop Products
IPAK Gard® pretreatment pack
Milli-Q® Lab Water Purification Consumables
Find the cartridges, UV lamps, filters and polishers for your water system. Advantages of using genuine Milli-Q consumables: guarantee water quality, sustainability features (new Drain Cap, Hg-free lamps). Designed for perfect fit.
Shop Products
Connect up to 4 POD dispensers to one Milli-Q® IQ water system
Accessories for Milli-Q® Lab Water Purification Systems
Milli-Q® water system accessories support lab-specific needs, improving safety, water quality, and productivity while conserving bench space.
Shop Products
CLRW-grade water supports accurate biomedical analyses
Purified water is a critical reagent in biomedical laboratories as it is essential for clinical assays and for rinsing and cleaning automated equipment. Any issues with water—such as contaminants, quality fluctuations or supply interruptions—can lead to clinical analyzer downtime, potential damage, and decreased laboratory efficiency. 
Appropriate water purification is crucial for maintaining laboratory operations and extending the lifespan of analyzers. The Clinical and Laboratory Standards Institute (CLSI) defines the quality of water required for clinical analyzers as Clinical Laboratory Reagent Water (CLRW).1 Using highly purified CLRW-grade water serves two main purposes: 
  • Supports accurate and reliable clinical analyses
  • Protects analyzers from contamination, clogging and damage
Impact of water contaminants on clinical assays
Consistent CLRW-quality water is vital for accurate and reliable assay results in clinical laboratories. Examples of water contaminants and their potential impacts on sensitive biomedical analyses include:
  • Inorganic ions (e.g. sodium, calcium, magnesium, chloride, etc.) can act as catalysts in organic and biochemical reactions.
  • Organic molecules may disturb biological experiments.
  • Bacteria and their by-products [e.g. alkaline phosphatase (ALP)]: Bacteria can contaminate samples and disrupt instrument operations; bacterial ALP may interfere with alkaline phosphatase used as a detection enzyme in immunoassays.
  • Particulates and colloids (e.g. silica, pollen, salts) can interfere with analyses, clog analyzers and reduce their lifespan.
Given the significant impact of water on clinical assays, laboratories should treat water as a critical reagent. Attention must be paid to water quality for each application. While clinical analyzers generally require CLRW-quality water, specific analyses (such as molecular biology, LC-MS, HPLC, or ICP-MS) may necessitate ultrapure water. Proper design, selection, and maintenance of water purification units can mitigate issues related to water quality, reduce instrument downtime, and enhance laboratory productivity and reliability.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents
1. Clinical and Laboratory Standards Institute (CLSI). Preparation and Testing of Reagent Water in the Medical Laboratory. 5th ed. CLSI guideline GP40. (ISBN 978-1-68440-266-3 [Print]; 978-1-68440-267-0 [Electronic]). Clinical and Laboratory Standards Institute, USA, 2024.