Skip to Content
Merck
All Photos(1)

Key Documents

04327

Sigma-Aldrich

Atto 612 Q

BioReagent, suitable for fluorescence

Synonym(s):

Atto 612 quencher, Atto Q 612

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352108
NACRES:
NA.32

Pricing and availability is not currently available.

product line

BioReagent

Assay

≥98.0% (HPCE)

manufacturer/tradename

ATTO-TEC GmbH

technique(s)

UV/Vis spectroscopy: suitable

solubility

DMF: soluble
DMSO: soluble

suitability

corresponds for UV test
suitable for fluorescence

storage temp.

−20°C

General description

Atto 612Q is a new quencher for fluorescence characterized by high molecular absorption (114.000). Atto 612Q is characterized by a high photostability and thermostability. It is moderately hydrophilic, and soluble in polar solvents as DMF or DMSO.

Application

Atto 612Q is a thiol reactive molecule making it an ideal candidate to modify cysteine residues [1].

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Valanne et al.
Analytical biochemistry, 375(1), 71-81 (2008-01-24)
The control of cell death is an intricate process involving a multitude of intracellular modulators. Among these molecules, the caspases have a central role and have become an interesting group of enzymes in the current pharmaceutical industry. We have developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service