Skip to Content
Merck
All Photos(1)

Key Documents

51028

Sigma-Aldrich

Atto 488 PPE

suitable for fluorescence

Synonym(s):

1-Palmitoyl-2-hydroxy-sn-glycero-3-phosphoethanolamine labeled with Atto 488

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

Assay

≥90% (TLC)

form

solid

manufacturer/tradename

ATTO-TEC GmbH

λ

in chloroform/methanol (8/2)

UV absorption

λ: 506-512 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 488 is a labeling dye with high molecular absorption (90,000) and quantum yield (0.80) as well as sufficient Stokes shift between excitation and emission maximum. It is optimized for excitation with an argon laser, and is characterized by high photostability.

Atto-Dye Labeled Phospholipids
Sigma-Aldrich offers a variety of glycero-phospholipids carrying one or two fatty acid groups (lipophilic groups) and a phosphate ester residue (hydrophilic group). They are labeled at the hydrophilic head group. After incorporation of the phospholipid into a membrane the fluorophore is located at the water/lipid interface of the membrane. We currently provide 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE), 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), palmitoyl-sn-glycero-phosphoethanolamine (PPE), and 1,2-dimyristoyl-snglycero-3-phospho-ethanolamine (DMPE) labeled with Atto-dyes.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Breaking the Diffraction Barrier in Fluorescence Microscopy by Optical Shelving.
Bretschneider, S.; Eggeling, Chr.; Hell, S. W.
Physical Review Letters, 98(21) (2007)
Dynamic Superresolution Imaging of Endogenous Proteins on Living Cells at Ultra-High Density.
Giannone, G.; et al.
Biophysical Journal, 99(4), 1303-1310 (2010)
Dual color localization microscopy of cellular nanostructures.
Gunkel, M.; et al.
Biotechnology Journal, 4(6), 927-938 (2009)
Flexible Microdomain Specific Staining of Block Copolymers for 3D Optical Nanoscopy.
Ullal, Chaitanya K.; et al.
Macromolecules, 44(19), 7508-7510 (2011)
Instant Live-Cell Super-Resolution Imaging of Cellular Structures by Nanoinjection of Fluorescent Probes.
Hennig, S.; et al.
Nano Letters, 15(2), 1374-1381 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service