Skip to Content
Merck
All Photos(1)

Key Documents

55607

Sigma-Aldrich

Atto 465 maleimide

BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Pricing and availability is not currently available.

product line

BioReagent

Assay

≥90% (coupling to thiols)

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 452 nm; λem 502 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

OCl(=O)(=O)=O.Nc1ccc2C=C3C=CC(=N)C=C3N(CCCC(=O)NCCN4C(=O)C=CC4=O)c2c1

InChI

1S/C23H23N5O3.ClHO4/c24-17-5-3-15-12-16-4-6-18(25)14-20(16)27(19(15)13-17)10-1-2-21(29)26-9-11-28-22(30)7-8-23(28)31;2-1(3,4)5/h3-8,12-14,24H,1-2,9-11,25H2,(H,26,29);(H,2,3,4,5)

InChI key

KHHWHZCVGMYUGL-UHFFFAOYSA-N

Application

Atto fluorescent labels are designed for high sensitivity applications, including single molecule detection. Atto labels have rigid structures that do not show any cis-trans-isomerization. Thus these labels display exceptional intensity with minimal spectral shift on conjugation. Atto 465 maleimide is suitable for fluorescence labeling of thiol/SH containing molecules.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radek Macháň et al.
Analytical and bioanalytical chemistry, 399(10), 3547-3554 (2011-02-05)
Z-scan fluorescence correlation spectroscopy (FCS) is employed to characterize the interaction between arenicin-1 and supported lipid bilayers (SLBs) of different compositions. Lipid analogue C8-BODIPY 500/510C5-HPC and ATTO 465 labelled arenicin-1 are used to detect changes in lipid and peptide diffusion
David J Posson et al.
Nature, 436(7052), 848-851 (2005-08-12)
Voltage-gated ion channels open and close in response to voltage changes across electrically excitable cell membranes. Voltage-gated potassium (Kv) channels are homotetramers with each subunit constructed from six transmembrane segments, S1-S6 (ref. 2). The voltage-sensing domain (segments S1-S4) contains charged
Thiol reactive probes and chemosensors.
Peng H, Chen W, Cheng Y, et al.
Sensors, 12, 15907-15946 (2012)
Waldemar Waldeck et al.
International journal of medical sciences, 8(2), 97-105 (2011-02-01)
Fluorescent proteins (FPs) are established tools for new applications, not-restricted to the cell biological research. They could also be ideal in surgery enhancing the precision to differentiate between the target tissue and the surrounding healthy tissue. FPs like the KillerRed
Gerald Donnert et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(31), 11440-11445 (2006-07-26)
We demonstrate far-field fluorescence microscopy with a focal-plane resolution of 15-20 nm in biological samples. The 10- to 12-fold multilateral increase in resolution below the diffraction barrier has been enabled by the elimination of molecular triplet state excitation as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service