Skip to Content
Merck
All Photos(1)

Key Documents

93259

Supelco

Atto 610-NHS ester

BioReagent, suitable for fluorescence, ≥90% (HPLC)

Synonym(s):

Atto 610 N-succinimidyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H34ClN3O8
Molecular Weight:
588.05
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Pricing and availability is not currently available.

product line

BioReagent

Assay

≥90% (HPLC)
≥90% (degree of coupling)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 613-619 nm Amax

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

SMILES string

[O-]Cl(=O)(=O)=O.C\[N+](C)=C1/C=CC2=Cc3cc4CCCN(CCCC(=O)ON5C(=O)CCC5=O)c4cc3C(C)(C)C2=C1

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica L Felhofer et al.
Electrophoresis, 31(15), 2469-2486 (2010-07-29)
Over the last years, there has been an explosion in the number of developments and applications of CE and microchip-CE. In part, this growth has been the direct consequence of recent developments in instrumentation associated with CE. This review, which
Lu Liu et al.
Protein science : a publication of the Protein Society, 16(11), 2403-2411 (2007-10-27)
Serpins inhibit serine proteases by mechanically disrupting the protease active site. The protease first reacts with the serpin's reactive center loop (RCL) to form an acylenzyme. Then the RCL inserts into a beta-sheet in the body of the serpin, translocating
A membrane-bound antiparallel dimer of rat islet amyloid polypeptide.
Abhinav Nath et al.
Angewandte Chemie (International ed. in English), 50(46), 10859-10862 (2011-09-29)
Xiao X Han et al.
Analytical chemistry, 80(17), 6567-6572 (2008-07-30)
A simple and effective protocol for detections of protein-protein and protein-small molecule interactions has been developed. After interactions between proteins and their corresponding ligands, we employed colloidal silver staining for producing active substrates for surface-enhanced Raman scattering (SERS) and surface-enhanced
Lu Liu et al.
Biochemistry, 45(36), 10865-10872 (2006-09-07)
Serpins regulate serine proteases by forming metastable covalent complexes with their targets. The protease docks with the serpin and cleaves the serpin's reactive center loop (RCL) forming an acylenzyme intermediate. Cleavage triggers insertion of the RCL into beta sheet A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service