Skip to Content
Merck
All Photos(1)

Key Documents

KSPRA2015

KiCqStart® Probe Assays

Predesigned probe assays for gene expression analysis

Synonym(s):

5’ nuclease assay, Hydrolysis Probes, Predesigned Probes, Probe-based Gene Expression, Probe-based RT-qPCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133

storage temp.

−20°C

General description

Overview
Predesigned KiCqStart Probe Assays are for quantifying gene expression by RT-qPCR (Reverse Transcription Quantitative Real-Time PCR). To comply with MIQE, they have been developed using sophisticated bioinformatics tools and validated in silico to avoid off-target amplification1. KiCqStart Probe Assays are available as up to three sets of a forward primer, a reverse primer, and a Dual-Labeled Probe for genes from common model organisms.

1SNP loci have been avoided during the design process when possible. Please consult dbSNP to determine if your gene has unavoidable SNPs.

To learn more about KiCqStart Probe Assays, visit our Frequently Asked Questions section.

Specifications
Design characteristics
  • GC Content: 20 - 80%2
  • Melting Temperature: 58.0 °C2
  • Secondary Structure: None
  • Excessive 3′ Primer Clamping: Minimal
  • Amplicon size: 75 - 150 bases
  • Splice variant detected: Most prevalent

Oligo characteristics (primers)
  • Molecule: DNA (all sequences verified by mass spec QC)
  • Internal Modifications: None
  • Contents: 1 forward and 1 reverse primer
  • Length: 18 - 27 bases2
  • Purification: RP cartridge
  • Format: 3 OD minimum yield, dry; 2 tubes with 1 oligonucleotide each

Oligo characteristics (probe)
  • Molecule: DNA (all sequences verified by mass spec QC)
  • Internal Modifications: None
  • 5′ Reporter Dye: 6-FAM or HEX

Application

Product use: gene expression analysis

Features and Benefits

KiCqStart® Probe Assays are sensitive, specific, and will save you time and money by eliminating the tedious steps required to develop an effective probe-based RT-qPCR Assay. The probe is labeled with Sigma′s dark Onyx Quencher (OQ), which emits heat instead of light and therefore offers an improved signal-to-noise ratio compared to traditional fluorescent quenchers.

Benefits of these assays include:
  • Target analysis is done
  • Design characteristics are set
  • Oligo characteristics are set
  • Easy to order

Physical properties

Properties include:
  • Available species: human, mouse, and rat
  • Components: three oligos, one per tube
  • Shipping condition: dry, ambient temperature
  • Storage condition: dry or wet, single-use aliquots

Other Notes

Frequently Asked Questions section is available for this product.

Legal Information

KiCqStart is a registered trademark of QIAGEN Beverly Inc.
OQ is a trademark of Sigma-Aldrich Co. LLC
Onyx Quencher is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service