A213
α2A-Adrenergic Receptor human
recombinant, expressed in baculovirus infected Sf9 cells, membrane suspension
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
recombinant
expressed in baculovirus infected Sf9 cells
form
membrane suspension
packaging
vial of 50 units
UniProt accession no.
storage temp.
−70°C
Gene Information
human ... ADRA2A(150)
Caution
Membranes retain original specific activity for several months when stored in original packaging solution.
Physical form
Suspension of cell membranes in 50 mM Tris-HCl, pH 7.4, containing 10% glycerol and 1% BSA.
Analysis Note
Recommended assay conditions are supplied on the product data sheet. This is a bioassayed product.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
A Renouard et al.
The Journal of pharmacology and experimental therapeutics, 270(3), 946-957 (1994-09-01)
In the present study, we examined the binding of the alpha-2 adrenergic receptor (AR) antagonist [3H]-(2-(2-methoxy-1,4-benzodioxan- 2yl)-2-imidazoline ([3H]RX821002) to alpha-2 AR in rat cerebral cortex (CC) and compared the properties of these sites to those of rat alpha-2A (R alpha-2A)
R G Lawhead et al.
Anesthesiology, 77(5), 983-991 (1992-11-01)
alpha-2 Adrenergic receptors can be subdivided into four subtypes based on their pharmacologic properties. The subtype of alpha-2 adrenergic receptor present in human spinal cord has not been reported previously. The affinities of nine alpha-2 subtype-selective drugs for the alpha-2
J W Lomasney et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(13), 5094-5098 (1990-07-01)
Pharmacologic, biochemical, and genetic analyses have demonstrated the existence of multiple alpha 2-adrenergic receptor (alpha 2AR) subtypes. We have cloned a human alpha 2AR by using the polymerase chain reaction with oligonucleotide primers homologous to conserved regions of the previously
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service