Skip to Content
Merck
All Photos(1)

Key Documents

E2643

Sigma-Aldrich

Enterostatin human

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H36N8O6
CAS Number:
Molecular Weight:
496.56
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

C[C@H](N)C(=O)N1CCC[C@H]1C(=O)NCC(=O)N2CCC[C@H]2C(=O)N[C@@H](CCCNC(N)=N)C(O)=O

InChI

1S/C21H36N8O6/c1-12(22)19(33)29-10-4-6-14(29)17(31)26-11-16(30)28-9-3-7-15(28)18(32)27-13(20(34)35)5-2-8-25-21(23)24/h12-15H,2-11,22H2,1H3,(H,26,31)(H,27,32)(H,34,35)(H4,23,24,25)/t12-,13-,14-,15-/m0/s1

InChI key

ITZMJCSORYKOSI-AJNGGQMLSA-N

Gene Information

human ... CLPS(1208)

Amino Acid Sequence

Ala-Pro-Gly-Pro-Arg

General description

Enterostatin is a pentapeptide Val-Pro-Asp-Pro-Arg (VPDPR) obtained from trypsin digestion of procolipase, which is a pancreatic lipase. It is produced by the intestine, and the gastric mucosa.

Application

Enterostatin human maybe used to study the hydrolytic activity of human angiotensin-converting enzyme-related carboxypeptidase (ACE2) on human biological peptides.

Biochem/physiol Actions

Peptide that inhibits fat intake in experiments involving dietary choice.
Studies show that peripheral and central injection of enterostatin reduces the food intake. In Sprague-Dawley rats, central injection leads to a reduction in the intake of high-fat food as opposed to low-fat food. Therefore, it might have a role in the feedback loop of dietary fat. In a chronic manner, it is responsible for the reduction of body fat, intake of fat and body weight. It controls the secretory activity of β-cells of pancreas, and suppresses insulin secretion without affecting the secretion of glucagon and somatostatin. It might also have anti-incretin activity in the enteroinsular axis, and might regulate postprandial hyperinsulinemia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R A Silvestre et al.
Diabetes, 45(9), 1157-1160 (1996-09-01)
Enterostatin is a pentapeptide generated by trypic digestion of procolipase in the small intestine. Both peripheral and central administration of this peptide to rats has been shown to reduce food intake, this reduction being due to specific suppression of fat
C Erlanson-Albertsson et al.
Obesity research, 5(4), 360-372 (1997-07-01)
A high fat intake, together with an inability to match lipid oxidation to fat intake, has been found to be correlated with obesity in humans. This review describes our current understanding of enterostatin, a peptide that selectively reduces fat intake.
C Erlanson-Albertsson et al.
Physiology & behavior, 49(6), 1191-1194 (1991-06-01)
Pancreatic procolipase is activated by trypsin forming colipase, a cofactor for pancreatic lipase involved in intestinal fat digestion and a pentapeptide named enterostatin. Enterostatin with the sequence Val-Pro-Asp-Pro-Arg (VPDPR) was previously shown to decrease food intake in rats both after
Chad Vickers et al.
The Journal of biological chemistry, 277(17), 14838-14843 (2002-01-30)
Human angiotensin-converting enzyme-related carboxypeptidase (ACE2) is a zinc metalloprotease whose closest homolog is angiotensin I-converting enzyme. To begin to elucidate the physiological role of ACE2, ACE2 was purified, and its catalytic activity was characterized. ACE2 proteolytic activity has a pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service