Skip to Content
Merck
All Photos(1)

Key Documents

L5170

Sigma-Aldrich

Leukotriene A4 Hydrolase human

recombinant, expressed in E. coli

Synonym(s):

LTA-4 hydrolase, LTA4, LTA4H

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

description

contains C-terminal his-tag

Assay

≥90% (SDS-PAGE)

mol wt

~69 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... LTA4H(4048)

Application

Leukotriene A4 Hydrolase human (LTA4H) is a drug target for anti-inflammation, and for cancer prevention and therapy. It is also suitable for screening inhibitors of Leukotriene B4 synthesis. LTA4H is used to study allergic asthma and airway hyperresponsiveness .

Biochem/physiol Actions

Leukotriene A4 Hydrolase human (LTA4H) is a bifunctional zinc metalloenzyme that converts LTA4 into Leukotriene B4, and also demonstrates aminopeptidase activity. Leukotriene B4 is a lipid chemoattractant that plays critical roles in inflammaton, immune responses, host defenses against infections, and lipid homeostasis. Inhibition of LTA4H in a mouse model decreases LTB4 in the airways and attenuates airway inflammation and airway hyperreactivity through modulation of T cell and dendritic cell function .

Physical form

Supplied as a solution in 100mM Tris, pH 8.0, containing 20% glycerol and 100mM potassium chloride.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Orning et al.
The Journal of biological chemistry, 269(15), 11269-11273 (1994-04-15)
Leukotriene-A4 hydrolase (EC 3.3.2.6) cleaved the NH2-terminal amino acid from several tripeptides, typified by arginyl-glycyl-aspartic acid, arginyl-glycyl-glycine, and arginyl-histidyl-phenylalanine, with catalytic efficiencies (kcat/Km) > or = 1 x 10(6) M-1 s-1. This exceeds by 10-fold the kcat/Km for its lipid
Mikell Paige et al.
Journal of immunology (Baltimore, Md. : 1950), 192(11), 5059-5068 (2014-04-29)
The leukotriene A4 hydrolase (LTA4H) is a bifunctional enzyme with epoxy hydrolase and aminopeptidase activities. We hypothesize that the LTA4H aminopeptidase activity alleviates neutrophilic inflammation, which contributes to cigarette smoke (CS)-induced emphysema by clearing proline-glycine-proline (PGP), a triamino acid chemokine
John W Newman et al.
Progress in lipid research, 44(1), 1-51 (2005-03-08)
The epoxide hydrolases (EHs) are enzymes present in all living organisms, which transform epoxide containing lipids by the addition of water. In plants and animals, many of these lipid substrates have potent biologically activities, such as host defenses, control of
J Haeggström et al.
The Journal of biological chemistry, 261(14), 6332-6337 (1986-05-15)
Mouse liver homogenates transformed leukotriene A4 into a 5,6-dihydroxy-7,9,11,14-eicosatetraenoic acid. This novel enzymatic metabolite of leukotriene A4 was characterized by physical means including ultraviolet spectroscopy, high performance liquid chromatography, and gas chromatography-mass spectrometry. After subcellular fractionation, the enzymatic activity was
M Minami et al.
The Journal of biological chemistry, 262(29), 13873-13876 (1987-10-15)
We have isolated a near full-length cDNA encoding human leukotriene A4 hydrolase, which synthesizes a potent chemotactic and spasmogenic compound, leukotriene B4. A human spleen cDNA library was screened with a 48-mer oligonucleotide probe, synthesized according to the partial amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service