Skip to Content
Merck
All Photos(1)

Key Documents

T6769

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O128:B12

trinitrophenyl conjugate

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352211

biological source

Escherichia coli (O128:B12)

conjugate

trinitrophenyl conjugate

Assay

≥98% (TLC)

form

powder

extent of labeling

15-60 μg trinitrophenolmg LPS

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kea Martin et al.
Journal of immunology (Baltimore, Md. : 1950), 203(11), 2791-2806 (2019-10-30)
The paracaspase Malt1 is a key regulator of canonical NF-κB activation downstream of multiple receptors in both immune and nonimmune cells. Genetic disruption of Malt1 protease function in mice and MALT1 mutations in humans results in reduced regulatory T cells
Jun Liu et al.
World journal of gastroenterology, 15(16), 2009-2015 (2009-04-29)
To investigate the effect of Lianshu preparation on lipopolysaccharide (LPS)-induced diarrhea in rats. A diarrhea model was established in Sprague Dawley rats via injection of 1 mL of 30 mg/kg LPS. A total of 40 rats were randomly divided into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service