MilliporeSigma
All Photos(6)
Key Documents
Z545481
Molymod® Individual organic model set
DNA (includes stand, 11cm H)
About This Item
UNSPSC Code:
60104506
NACRES:
NB.17
description
base-pairs 2
manufacturer/tradename
Molymod
General description
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Legal Information
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.