Key Documents
Z548820
Sigma-Aldrich
Aldrich® solvent-pouring funnel
stem L × O.D. 100 mm × 22 mm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
stem L × O.D.
100 mm × 22 mm
General description
Unique, closed-end glass funnel has 2 mm diameter holes in the stem permitting the gentle addition of solvents to chromatography columns without disturbing the silica gel/sand layer. The top of the funnel is 75 mm in diameter.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC