Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

62501-046

Sigma-Aldrich

DNAgard® Blood

greener alternative

100mL bottle, for room temp preservation of DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

grade

for molecular biology

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

room temp

General description

DNAgard Blood is designed for the immediate stabilization of DNA in blood with the convenience of room temperature shipping and storage. The preservative reagent rapidly permeates cellular structures and membranes to stabilize and protect genomic DNA in whole blood samples. The use of DNAgard Blood allows a streamlined workflow from blood collection in the clinic or field to sample processing in the laboratory.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product was Designed for Energy Efficiency. To see how Stanford incorporated Biomatrica Technology and saved substantial amounts of energy, see their published study:

Biomatrica technology enables safe storage of biological material at room temperature.

Packaging

1 bottle

Physical form

DNAgard Blood reagent 100 mL (stabilizes at least 400 mL of blood)

Legal Information

DNAgard is a registered trademark of Biomatrica

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malini Udtha et al.
Biopreservation and biobanking, 12(5), 332-336 (2014-10-24)
Preservation of biospecimens for biobanking applications traditionally involves freezing while maintaining the integrity of the product throughout multiple freeze-thaw cycles. The protection and stabilization of DNA at room temperature (RT) may eliminate the costs associated with freezer storage and reduce

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service