Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z707392

SealPlate® MiniStrips film

sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

polyester film

sterility

sterile

feature

piercable

packaging

pack of 200 ea

manufacturer/tradename

Excel Scientific SPS-2X8-50

technique(s)

ELISA: suitable

L

102.9 mm

W × L

1.9 cm × 10.29 cm , 8.26 cm with Tabs removed

W

82.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

MiniStrips films are identical to SealPlate films, but designed for sealing one or two 8 well rows at a time. They may be used on strip-well plates or standard plates whenever rows must be selectively protected or accessed. Use as a secondary seal to cover films that have already been pierced.

Legal Information

MiniStrips is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service