Skip to Content
MilliporeSigma
(±)-Citronellal analytical standard

(±)-Citronellal

(±)-Citronellal analytical standard
Synonym(s):
(±)-3,7-Dimethyl-6-octenal
Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing