Skip to Content
MilliporeSigma
(±)-Propylene oxide analytical standard

(±)-Propylene oxide

(±)-Propylene oxide analytical standard
Synonym(s):
(±)-Methyloxirane, 1,2-Epoxypropane
Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3