Skip to Content
MilliporeSigma
Dodecanoic acid analytical standard

Dodecanoic acid

Dodecanoic acid analytical standard
Synonym(s):
ABL, Lauric acid, NSC 5026
Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing