Skip to Content
MilliporeSigma
γ-Aminobutyric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

γ-Aminobutyric acid

γ-Aminobutyric acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
3-Carboxypropylamine, 4-Aminobutanoic acid, GABA, Piperidic acid, Piperidinic acid
Linear Formula:
NH2(CH2)3COOH
CAS Number:
Molecular Weight:
103.12
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Analyte Suitability
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique