Skip to Content
MilliporeSigma
vibrant-m

Human Insulin in PBS solution

vibrant-m
Synonym(s):
Human Insulin solution
Empirical Formula (Hill Notation):
C257H383N65O77S6
CAS Number:
Molecular Weight:
5807.57

Products

Brand
Feature
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade