Skip to Content
MilliporeSigma
Linuron-(methyl-d3, methoxy-d3) PESTANAL®, analytical standard

Linuron-(methyl-d3, methoxy-d3)

Linuron-(methyl-d3, methoxy-d3) PESTANAL®, analytical standard
Synonym(s):
3-(3,4-Dichlorophenyl)-1-methoxy-d3-1-methyl-d3-urea, Linuron-d6
Empirical Formula (Hill Notation):
C9D6H4Cl2N2O2
CAS Number:
Molecular Weight:
255.13

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing