Skip to Content
MilliporeSigma
N1-Acetyl-N2-formyl-5-methoxykynuramine ≥95% (HPLC)

N1-Acetyl-N2-formyl-5-methoxykynuramine

N1-Acetyl-N2-formyl-5-methoxykynuramine ≥95% (HPLC)
Synonym(s):
AFMK, N-[3-[2-(Formylamino)-5-methoxyphenyl]-3-oxopropyl]acetamide, NSC 688263
Empirical Formula (Hill Notation):
C13H16N2O4
CAS Number:
Molecular Weight:
264.28

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing