
NA2F Glycan

Synonym(s):
(Gal)2(GlcNAc)2(Man)3(GlcNAc)2Fuc, Asialo, galactosylated, biantennary N-glycan, core fucosylated, Mannotriose-[fucosyl-di-(N-acetylglucosamine)], bis(galactosyl-N-acetylglucosaminyl)
Empirical Formula (Hill Notation):
C68H114N4O50
CAS Number:
Molecular Weight:
1787.63
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1