
Palonosetron Related Compound B

Synonym(s):
(3aS)-2,4,5,6-Tetrahydro-2-[(3S)-1-oxido-1-azabicyclo[2.2.2]oct-3-yl]-1H-benz[de]isoquinolin-1-one
Empirical Formula (Hill Notation):
C19H22N2O2
CAS Number:
Molecular Weight:
310.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing