Proguanil for system suitability
Synonym(s):
Proguanil hydrochloride, Chlorguanide, N1-(4-Chlorophenyl)-N5-isopropylbiguanide
Empirical Formula (Hill Notation):
C11H16ClN5·HCl
CAS Number:
Molecular Weight:
290.19
Products
Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing