MilliporeSigma
  • A Requirement for Zic2 in the Regulation of Nodal Expression Underlies the Establishment of Left-Sided Identity.
A Requirement for Zic2 in the Regulation of Nodal Expression Underlies the Establishment of Left-Sided Identity.
Scientific reports (2018-07-12)
Iain M Dykes, Dorota Szumska, Linta Kuncheria, Rathi Puliyadi, Chiann-Mun Chen, Costis Papanayotou, Helen Lockstone, Christèle Dubourg, Véronique David, Jurgen E Schneider, Thomas M Keane, David J Adams, Steve D M Brown, Sandra Mercier, Sylvie Odent, Jérôme Collignon, Shoumo Bhattacharya
ABSTRACT
ZIC2 mutation is known to cause holoprosencephaly (HPE). A subset of ZIC2 HPE probands harbour cardiovascular and visceral anomalies suggestive of laterality defects. 3D-imaging of novel mouse Zic2 mutants uncovers, in addition to HPE, laterality defects in lungs, heart, vasculature and viscera. A strong bias towards right isomerism indicates a failure to establish left identity in the lateral plate mesoderm (LPM), a phenotype that cannot be explained simply by the defective ciliogenesis previously noted in Zic2 mutants. Gene expression analysis showed that the left-determining NODAL-dependent signalling cascade fails to be activated in the LPM, and that the expression of Nodal at the node, which normally triggers this event, is itself defective in these embryos. Analysis of ChiP-seq data, in vitro transcriptional assays and mutagenesis reveals a requirement for a low-affinity ZIC2 binding site for the activation of the Nodal enhancer HBE, which is normally active in node precursor cells. These data show that ZIC2 is required for correct Nodal expression at the node and suggest a model in which ZIC2 acts at different levels to establish LR asymmetry, promoting both the production of the signal that induces left side identity and the morphogenesis of the cilia that bias its distribution.
Sigma-Aldrich
Hanks′ Balanced Salt Solution 10x, Without calcium chloride, magnesium sulfate and sodium bicarbonate, 10 ×, liquid, sterile-filtered, suitable for cell culture