MilliporeSigma
  • A Re(V)-catalyzed C-N bond-forming route to human lipoxygenase inhibitors.
A Re(V)-catalyzed C-N bond-forming route to human lipoxygenase inhibitors.
Organic letters (2005-06-04)
Rachana V Ohri, Alexander T Radosevich, K James Hrovat, Christine Musich, David Huang, Theodore R Holman, F Dean Toste
ABSTRACT
[reaction: see text] A regioselective synthesis of propargylamines by the coupling of propargyl alcohols with tosylamines and carbamates catalyzed by an air- and moisture-tolerant rhenium-oxo complex is described. The ability to couple functionalized components allows for convergent approaches to nitrogen-containing heterocyclic compounds such as the marine antibiotic pentabromopseudilin. These compounds were assayed against human lipoxygenase and found to be both potent and selective.
Sigma-Aldrich
Pentabromopseudilin, ≥97% (HPLC)