MilliporeSigma
  • 27-hydroxycholesterol induces production of tumor necrosis factor-alpha from macrophages.
27-hydroxycholesterol induces production of tumor necrosis factor-alpha from macrophages.
Biochemical and biophysical research communications (2012-12-19)
Sun-Mi Kim, Hyojung Jang, Yonghae Son, Sae-A Lee, Sun-Sik Bae, Young Chul Park, Seong-Kug Eo, Koanhoi Kim
ABSTRACT
Enhanced production of TNF-α from macrophages promotes development and instability of atherosclerotic plaques, but involvement of lipid component in TNF-α production has not been clarified in atherosclerosis. We attempted to determine whether cholesterol oxidation products (oxysterols) could modify TNF-α production. Treatment of THP-1 cells with 27-hydroxycholesterol (27OHChol) or 7α-hydroxycholesterol (7αOHChol) resulted in a profound increase in TNF-α transcription, while treatment with an identical concentration of cholesterol and 7-ketochoelsterol did not lead to any change in TNF-α expression. Treatment with 27OHChol resulted in increased synthesis, as well as secretion, of TNF-α, while 7αOHChol led to increased synthesis of TNF-α without affecting secretion of the cytokine. Co-treatment with 7αOHChol or 27OHChol and LPS resulted in synergistically enhanced or augmented secretion of TNF-α. Treatment with TO-901317, pertussis toxin, PP2, and LY294002 resulted not only in attenuated transcription of TNF-α induced by 27OHChol and 7αOHChol, but also secretion of TNF-α enhanced by 27OHChol. This is the first report demonstrating enhanced production of TNF-α in macrophages by treatment with oxysterols which are detected in abundance in atheromatous lesions; in addition, results of the current study provide evidence indicating that certain types of oxysterols contribute to development of atherosclerosis by promoting production of proinflammatory cytokines.
Product Description
Avanti
27-hydroxycholesterol, Avanti Research - A Croda Brand