MilliporeSigma
  • Characterization of Cfa1, a monofunctional acyl carrier protein involved in the biosynthesis of the phytotoxin coronatine.
Characterization of Cfa1, a monofunctional acyl carrier protein involved in the biosynthesis of the phytotoxin coronatine.
Journal of bacteriology (2004-04-03)
Heather Seidle, Vidhya Rangaswamy, Robin Couch, Carol L Bender, Ronald J Parry
ABSTRACT
Cfa1 was overproduced in Escherichia coli and Pseudomonas syringae, and the degree of 4'-phosphopantetheinylation was determined. The malonyl-coenzyme A:acyl carrier protein transacylase (FabD) of P. syringae was overproduced and shown to catalyze malonylation of Cfa1, suggesting that FabD plays a role in coronatine biosynthesis. Highly purified Cfa1 did not exhibit self-malonylation activity.
Sigma-Aldrich
Malonyl coenzyme A tetralithium salt, ≥90% (HPLC)