MilliporeSigma
  • Molecular modeling and dynamics studies of cytidylate kinase from Mycobacterium tuberculosis H37Rv.
Molecular modeling and dynamics studies of cytidylate kinase from Mycobacterium tuberculosis H37Rv.
Journal of molecular modeling (2008-03-18)
Rafael Andrade Caceres, Luís Fernando Saraiva Macedo Timmers, Ana Luiza Vivan, Cristopher Zandoná Schneider, Luiz Augusto Basso, Walter Filgueira De Azevedo, Diogenes Santiago Santos
ABSTRACT
Bacterial cytidylate kinase or cytidine monophosphate kinase (CMP kinase) catalyses the phosphoryl transfer from ATP to CMP and dCMP, resulting in the formation nucleoside diphosphates. In eukaryotes, CMP/UMP kinase catalyses the conversion of UMP and CMP to, respectively, UDP and CDP with high efficiency. This work describes for the first time a model of bacterial cytidylate kinase or cytidine monophosphate kinase (CMP kinase) from mycobacterium tuberculosis (MtCMPK). We modeled MtPCMPK in apo form and in complex with cytidine 5'-monophosphate (CMP) to try to determine the structural basis for specificity. Comparative analysis of the model of MtCMPK allowed identification of structural features responsible for ligand affinities. Analysis of the molecular dynamics simulations of these two systems indicates the structural features responsible for the stability of the structure, and may help in the identification of new inhibitors for this enzyme.
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate sodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
2′-Deoxycytidine 5′-monophosphate, Sigma Grade, ≥95.0%