MilliporeSigma
  • Matrix metalloproteinase-9 silencing by RNA interference triggers the migratory-adhesive switch in Ewing's sarcoma cells.
Matrix metalloproteinase-9 silencing by RNA interference triggers the migratory-adhesive switch in Ewing's sarcoma cells.
The Journal of biological chemistry (2003-07-09)
Josiane Sancéau, Sandrine Truchet, Brigitte Bauvois
ABSTRACT
Enhanced expression of (pro)matrix metalloproteinase-9 (MMP-9) is associated with human tumor invasion and/or metastasis. COH cells derived from a highly invasive and metastatic Ewing's sarcoma constitutively express proMMP-9. Transfection of a double stranded RNA that targets the MMP-9 mRNA into COH cells depleted the corresponding mRNA and protein as demonstrated by reverse transcriptase-PCR, enzyme-linked immunosorbent assay, and gelatin zymography. proMMP-9 extinction resulted in the following: (i) decreased spreading on extracellular matrix (fibronectin, laminin, collagen IV)-coated surfaces, (ii) inhibition of migration toward fibronectin, and (iii) induced aggregation, which was specifically disrupted by a function-blocking E-cadherin antibody. MMP-9 knockdown concomitantly resulted in increased levels of surface E-cadherin, redistribution at the plasma membrane of beta-catenin, and its physical association with E-cadherin. Moreover, induction of E-cadherin-mediated adhesion was associated with RhoA activation and changes in paxillin cytoskeleton. Finally, an inhibitor of gelatinolytic activity of pro-MMP9 did not reduce COH cell migration confirming that the enzymatic property of COH MMP-9 was not required for migration toward fibronectin. Overall, our observations define a novel critical role for proMMP-9 in providing a cellular switch between stationary and migratory cell phases.