MilliporeSigma
Biochemical properties of polysaccharides from black pepper.
Biological & pharmaceutical bulletin (2002-09-17)
Hyug Chun, Dong Hoon Shin, Bum Shik Hong, Won Dai Cho, Hong Yon Cho, Han Chul Yang
ABSTRACT
The purified polysaccharides from Piper nigrum were prepared as follows: a hot water extract of pepper seeds was fractionated by ultrafiltration with a 5-kDa-membrane cartridge. A fraction with 5 kDa or bigger molecules was successively purified by open column chromatography on DEAE-Toyopearl 650C and Bio-gel P-60 with each active fraction, resulting in PN-Ib and PN-IIa, purified anti-complementary polysaccharides. None of the anti-complementary activity of any polysaccharide was changed by pronase digestion or polymyxin B treatment, but they were decreased by periodate oxidation. Analysis of component sugar and molecular mass determination of the anti-complementary polysaccharides indicated that PN-Ib with an average molecular mass of 21 kDa contained 88.5% glucose and other negligible minor monosaccharides, while PN-IIa showed a different monosaccharide composition, which contained a significant proportion of galactose, arabinose, galacturonic acid and rhamnose. The molar ratio of galactose and arabinose of PN-IIa (48 kDa) was 1.93:1. PN-1 did not react with beta-glucosyl Yariv reagent, however, PN-IIa did react, which indicated that PN-IIa might be an arabinogalactan. Based upon these results, the usefulness of purified anti-complementary polysaccharides from Piper nigrum is suggested as a supplement for immune enhancement.
Product Description
TOYOPEARL® DEAE-650C Bulk Media, phase DEAE (diethylaminoethyl), bottle of 250 mL, 100 μm particle size