MilliporeSigma
  • Conjugate Addition-Enantioselective Protonation of N-Aryl Glycines to α-Branched 2-Vinylazaarenes via Cooperative Photoredox and Asymmetric Catalysis.
Conjugate Addition-Enantioselective Protonation of N-Aryl Glycines to α-Branched 2-Vinylazaarenes via Cooperative Photoredox and Asymmetric Catalysis.
Journal of the American Chemical Society (2018-04-11)
Yanli Yin, Yating Dai, Hongshao Jia, Jiangtao Li, Liwei Bu, Baokun Qiao, Xiaowei Zhao, Zhiyong Jiang
ABSTRACT
An enantioselective protonation strategy has been successfully applied to the synthesis of chiral α-tertiary azaarenes. With a dual catalytic system involving a chiral phosphoric acid and a dicyanopyrazine-derived chromophore (DPZ) photosensitizer that is mediated by visible light, a variety of α-branched 2-vinylpyridines and 2-vinylquinolines with N-aryl glycines underwent a redox-neutral, radical conjugate addition-protonation process and provided valuable chiral 3-(2-pyridine/quinoline)-3-substituted amines in high yields with good to excellent enantioselectivities (up to >99% ee). An application of this methodology to a two-step synthesis of the enantiomerically pure medicinal compound pheniramine (Avil) is also presented.
Sigma-Aldrich
DPZ, 95%