MilliporeSigma
  • Omics-based analyses revealed metabolic responses of Clostridium acetobutylicum to lignocellulose-derived inhibitors furfural, formic acid and phenol stress for butanol fermentation.
Omics-based analyses revealed metabolic responses of Clostridium acetobutylicum to lignocellulose-derived inhibitors furfural, formic acid and phenol stress for butanol fermentation.
Biotechnology for biofuels (2019-05-07)
Huanhuan Liu, Jing Zhang, Jian Yuan, Xiaolong Jiang, Lingyan Jiang, Guang Zhao, Di Huang, Bin Liu
ABSTRACT
Clostridium acetobutylicum is a model fermentative anaerobe for consolidated bioprocessing of lignocellulose hydrolysates into acetone-butanol-ethanol (ABE). However, the main inhibitors (acids, furans and phenols) ubiquitous in lignocellulose hydrolysates strictly limit the conversion efficiency. Thus, it is essential to understand the underlying mechanisms of lignocellulose hydrolysate inhibitors to identify key industrial bottlenecks that undermine efficient biofuel production. The recently developed omics strategy for intracellular metabolites and protein quantification now allow for the in-depth mapping of strain metabolism and thereby enable the resolution of the underlying mechanisms. The toxicity of the main inhibitors in lignocellulose hydrolysates against C. acetobutylicum and ABE production was systematically investigated, and the changes in intracellular metabolism were analyzed by metabolomics and proteomics. The toxicity of the main lignocellulose hydrolysate inhibitors at the same dose was ranked as follows: formic acid > phenol > furfural. Metabolomic analysis based on weighted gene coexpression network analysis (WGCNA) revealed that the three inhibitors triggered the stringent response of C. acetobutylicum. Proteomic analysis based on peptide mass spectrometry (MS) supported the above results and provided more comprehensive conclusions. Under the stress of three inhibitors, the metabolites and key enzymes/proteins involved in glycolysis, reductive tricarboxylic acid (TCA) cycle, acetone-butanol synthesis and redox metabolism were lower than those in the control group. Moreover, proteins involved in gluconeogenesis, the oxidative TCA cycle, thiol peroxidase (TPX) for oxidative stress were significantly upregulated, indicating that inhibitor stress induced the stress response and metabolic regulation. In addition, the three inhibitors also showed stress specificity related to fatty acid synthesis, ATP synthesis, nucleic acid metabolism, nicotinic acid metabolism, cell wall synthesis, spore synthesis and flagellum synthesis and so on. Integrated omics platforms provide insight into the cellular responses of C. acetobutylicum to cytotoxic inhibitors released during the deconstruction of lignocellulose. This insight allows us to fully improve the strain to adapt to a challenging culture environment, which will prove critical to the industrial efficacy of C. acetobutylicum.