MilliporeSigma
  • Inhibition of Inflammation-Associated Thrombosis with ROS-Responsive Heparin-DOCA/PVAX Nanoparticles.
Inhibition of Inflammation-Associated Thrombosis with ROS-Responsive Heparin-DOCA/PVAX Nanoparticles.
Macromolecular bioscience (2019-06-22)
Zehong Xiang, Yanming Wang, Zhifang Ma, Zhirong Xin, Runhai Chen, Qiang Shi, Shing-Chung Wong, Jinghua Yin
ABSTRACT
Inflammation-associated thrombosis is a non-negligible source of mortalities and morbidities worldwide. To manipulate inflammation-associated coagulation, nanoparticles that contain anti-inflammatory polymer (copolyoxalate containing vanillyl alcohol, PVAX) and anti-thrombotic heparin derivative deoxycholic acid (Hep-DOCA) are prepared. The strategy takes advantage of the reducted side effects of heparin through heparin conjugation, achievement of long-term anti-inflammation by inflammation-trigged release of anti-inflammatory agents, and formation of PVAX/heparin-DOCA nanoparticles by co-self-assembly. It is demonstrated that the Hep-DOCA conjugate and PVAX are synthesized successfully; PVAX and Hep-DOCA nanodrugs (HDP) are obtained by co-assembly; the HDP nanoparticles effectively reduce the inflammation and coagulation without inducing lethal bleeding both in vivo and in vitro. The method provided here is versatile and effective, which paves new way to develop nanodrugs to treat inflammation-associated thrombosis safely.