MilliporeSigma
  • LncRNA MIR4435-2HG is downregulated in osteoarthritis and regulates chondrocyte cell proliferation and apoptosis.
LncRNA MIR4435-2HG is downregulated in osteoarthritis and regulates chondrocyte cell proliferation and apoptosis.
Journal of orthopaedic surgery and research (2019-08-08)
Yu Xiao, Yucheng Bao, Liang Tang, Li Wang
ABSTRACT
MIR4435-2HG is an oncogenic lncRNA in gastric cancer and lung cancer. Our preliminary microarray data showed that MIR4435-2HG was downregulated in osteoarthritis plasma specimen, indicating the possible involvement of MIR4435-2HG in osteoarthritis. MIR4435-2HG was downregulated in plasma of osteoarthritis than in plasma of healthy controls. Reduced levels of MIR4435-2HG expression effectively distinguished osteoarthritis patients from the control group. Expression levels of MIR4435-2HG increased after treatment. Overexpression of MIR4435-2HG promoted, while MIR4435-2HG knockdown inhibited the proliferation of chondrocytes. In contrast, MIR4435-2HG overexpression inhibited, while MIR4435-2HG knockdown promoted the apoptosis of chondrocytes. MIR4435-2HG is downregulated in osteoarthritis and regulates chondrocyte cell proliferation and apoptosis.