MilliporeSigma
  • Synergistic effects of ultrasound and photodynamic therapy leading to biofilm eradication on polyurethane catheter surfaces modified with hypericin nanoformulations.
Synergistic effects of ultrasound and photodynamic therapy leading to biofilm eradication on polyurethane catheter surfaces modified with hypericin nanoformulations.
Materials science & engineering. C, Materials for biological applications (2019-07-28)
Hendrik Vögeling, Nikola Plenagl, Benjamin Sebastian Seitz, Lili Duse, Shashank Reddy Pinnapireddy, Eyas Dayyoub, Jarmila Jedelska, Jana Brüßler, Udo Bakowsky
ABSTRACT
Catheter related infections are causing one third of all blood stream infections. The mortality of those infections is very high and the gold standard for catheter related blood stream infections (CR-BSI) is still the removal of the catheter and systemic antibiotic therapy. There already exist some approaches to prevent the biofilm formation on catheter material, which are far from ideal. A new strategy to prevent bacterial colonization on catheter surfaces is the application of photodynamic therapy (PDT). Therefor the surface has to be modified with substances that can be activated by light, leading to the production of cell toxic reactive oxygen species (ROS). Only small concentrations of the so called photosensitizer (PS) are necessary, avoiding side effects in human therapy. Furthermore, there is no resistance development in PDT. In this study polyurethane (PUR) surfaces were coated with hypericin nanoformulations, leading to 4.3 log10 reduction in bacterial growth in vitro. The effect could be enhanced by the application of ultrasound. The combination of PDT with ultrasound therapy led to a synergistic effect resulting in a 6.8 log10 reduction of viable counts. This minimal invasive method requires only an optical fibre inserted in the catheter lumen and an ultrasound device. Thus the implementation in daily clinical practice is very simple.