MilliporeSigma
Dextran-Coated Zinc-Doped Hydroxyapatite for Biomedical Applications.
Polymers (2019-05-18)
Daniela Predoi, Simona Liliana Iconaru, Mihai Valentin Predoi
ABSTRACT
Dextran-coated zinc-doped hydroxyapatite (ZnHApD) was synthesized by an adapted sol-gel method. The stability of ZnHApD nanoparticles in an aqueous solution was analyzed using ultrasonic measurements. The analysis of the evolution in time of the attenuation for each of the frequencies was performed. The X-ray diffraction (XRD) investigations exhibited that no impurity was found. The morphology, size and size distribution of the ZnHApD sample was investigated by transmission electron microscopy (TEM) and scanning electron microscopy (SEM). The TEM and SEM results showed that the ZnHApD particles have an ellipsoidal shape and a narrow distribution of sizes. The cell growth and toxicity of HEK-293 cells were investigated on the ZnHApD solution for four different concentrations and analyzed after 24 and 48 h. The ZnHApD solution presented a non-toxic activity against HEK-293 cells for all analyzed concentrations. The antibacterial assay revealed that all the tested microorganisms were inhibited by the ZnHApD dispersion after 24 and 48 h of incubation. It was observed that the effect of the ZnHApD solution on bacteria growth depended on the bacterial strain. The Porphyromonas gingivalis ATCC 33277 bacterial strain was the most sensitive, as a growth inhibition in the presence of 0.075 μg/mL ZnHApD in the culture medium was observed.