MilliporeSigma
  • Field-evolved resistance of Helicoverpa zea (Boddie) to transgenic maize expressing pyramided Cry1A.105/Cry2Ab2 proteins in northeast Louisiana, the United States.
Field-evolved resistance of Helicoverpa zea (Boddie) to transgenic maize expressing pyramided Cry1A.105/Cry2Ab2 proteins in northeast Louisiana, the United States.
Journal of invertebrate pathology (2019-03-03)
Gagandeep Kaur, Jianguo Guo, Sebe Brown, Graham P Head, Paula A Price, Silvana Paula-Moraes, Xinzhi Ni, Marcelo Dimase, Fangneng Huang
ABSTRACT
The corn earworm, Helicoverpa zea (Boddie) (Lepidoptera: Noctuidae), is a major target pest of pyramided Bt maize and cotton in the U.S. In 2017 and 2018, notable ear damage and larval survival of H. zea were observed on pyramided Cry1A.105/Cry2Ab2 maize in some fields in northeast Louisiana, U.S. The objective of this study was to determine if the field control problem was due to resistance development to the Bt proteins in plants. A total of 15 H. zea populations were collected from Bt and non-Bt maize plants in 2017 and 2018 in multiple locations in Louisiana, Florida, and Georgia. Diet-overlay bioassays showed that LC50s of Cry1A.105 and Cry2Ab2 for populations collected from the areas with control problems in northeast Louisiana were as much as >1623- and 88-fold greater than that of a susceptible strain, respectively. In addition, two field trials in 2018 validated that Cry1A.105/Cry2Ab2 maize failed in managing natural H. zea populations, while Bt maize containing Vip3A was effective in northeast Louisiana. Results of the study documented that the observed field control problems of Cry1A.105/Cry2Ab2 maize against H. zea in northeast Louisiana were due to resistance development of the insect to the Bt proteins in plants. This is the first documentation of field-evolved resistance to pyramided Bt maize in a target insect species in southern U.S. However, susceptibility levels to Cry1A.105 and Cry2Ab2 varied greatly among populations collected from the three states, suggesting uneven distributions of the resistance in the region.