MilliporeSigma
  • Redox-Active Ferrocene grafted on H-Terminated Si(111): Electrochemical Characterization of the Charge Transport Mechanism and Dynamics.
Redox-Active Ferrocene grafted on H-Terminated Si(111): Electrochemical Characterization of the Charge Transport Mechanism and Dynamics.
Scientific reports (2019-06-21)
Claudio Fontanesi, Enrico Da Como, Davide Vanossi, Monica Montecchi, Maria Cannio, Prakash Chandra Mondal, Walter Giurlani, Massimo Innocenti, Luca Pasquali
ABSTRACT
Electroactive self-assembled monolayers (SAMs) bearing a ferrocene (Fc) redox couple were chemically assembled on H-terminated semiconducting degenerate-doped n-type Si(111) substrate. This allows to create a Si(111)|organic-spacer|Fc hybrid interface, where the ferrocene moiety is covalently immobilized on the silicon, via two alkyl molecular spacers of different length. Organic monolayer formation was probed by Laser Ablation-Inductively Coupled Plasma-Mass Spectrometry (LA-ICP-MS) and X-ray photoelectron spectroscopy (XPS) measurements, which were also used to estimate thickness and surface assembled monolayer (SAM) surface coverage. Atomic force microscopy (AFM) measurements allowed to ascertain surface morphology and roughness. The single electron transfer process, between the ferrocene redox probe and the Si electrode surface, was probed by cyclic voltammetry (CV) measurements. CVs recorded at different scan rates, in the 10 to 500 mV s-1 range, allowed to determine peak-to-peak separation, half-wave potential, and charge-transfer rate constant (KET). The experimental findings suggest that the electron transfer is a one electron quasi-reversible process. The present demonstration of surface engineering of functional redox-active organometallic molecule can be efficient in the field of molecular electronics, surface-base redox chemistry, opto-electronic applications.