MilliporeSigma
  • CdsA is involved in biosynthesis of glycolipid MPIase essential for membrane protein integration in vivo.
CdsA is involved in biosynthesis of glycolipid MPIase essential for membrane protein integration in vivo.
Scientific reports (2019-02-06)
Katsuhiro Sawasato, Ryo Sato, Hanako Nishikawa, Naoki Iimura, Yuki Kamemoto, Kohki Fujikawa, Toshiyuki Yamaguchi, Yutetsu Kuruma, Yasushi Tamura, Toshiya Endo, Takuya Ueda, Keiko Shimamoto, Ken-Ichi Nishiyama
ABSTRACT
MPIase is a glycolipid that is involved in membrane protein integration. Despite evaluation of its functions in vitro, the lack of information on MPIase biosynthesis hampered verification of its involvement in vivo. In this study, we found that depletion of CdsA, a CDP-diacylglycerol synthase, caused not only a defect in phospholipid biosynthesis but also MPIase depletion with accumulation of the precursors of both membrane protein M13 coat protein and secretory protein OmpA. Yeast Tam41p, a mitochondrial CDP-diacylglycerol synthase, suppressed the defect in phospholipid biosynthesis, but restored neither MPIase biosynthesis, precursor processing, nor cell growth, indicating that MPIase is essential for membrane protein integration and therefore for cell growth. Consistently, we observed a severe defect in protein integration into MPIase-depleted membrane vesicles in vitro. Thus, the function of MPIase as a factor involved in protein integration was proven in vivo as well as in vitro. Moreover, Cds1p, a eukaryotic CdsA homologue, showed a potential for MPIase biosynthesis. From these results, we speculate the presence of a eukaryotic MPIase homologue.