MilliporeSigma
  • Bulk phase behavior of lithium imide-metal nitride ammonia decomposition catalysts.
Bulk phase behavior of lithium imide-metal nitride ammonia decomposition catalysts.
Physical chemistry chemical physics : PCCP (2018-08-24)
Joshua W Makepeace, Thomas J Wood, Phillip L Marks, Ronald I Smith, Claire A Murray, William I F David
ABSTRACT
Lithium imide is a promising new catalyst for the production of hydrogen from ammonia. Its catalytic activity has been reported to be significantly enhanced through its use as a composite with various transition metal nitrides. In this work, two of these composite catalysts (with manganese nitride and iron nitride) were examined using in situ neutron and X-ray powder diffraction experiments in order to explore the bulk phases present during ammonia decomposition. Under such conditions, the iron composite was found to be a mixture of lithium imide and iron metal, while the manganese composite contained lithium imide and manganese nitride at low temperatures, and a mixture of lithium imide and two ternary lithium-manganese nitrides (LixMn2-xN and a small proportion of Li7MnN4) at higher temperatures. The results indicate that the bulk formation of a ternary nitride is not necessary for ammonia decomposition in lithium imide-transition metal catalyst systems.