MilliporeSigma
  • The Clinical Significance of Programmed Death-1, Regulatory T Cells and Myeloid Derived Suppressor Cells in Patients with Nontuberculous Mycobacteria-Lung Disease.
The Clinical Significance of Programmed Death-1, Regulatory T Cells and Myeloid Derived Suppressor Cells in Patients with Nontuberculous Mycobacteria-Lung Disease.
Journal of clinical medicine (2019-05-28)
Chin-Chung Shu, Sheng-Wei Pan, Jia-Yih Feng, Jann-Yuan Wang, Yu-Jiun Chan, Chong-Jen Yu, Wei-Juin Su
ABSTRACT
Increasing expression of programmed death-1 (PD-1) in patients with nontuberculous mycobacteria lung disease (NTM-LD) has been reported, but its role in clinical characteristics and outcomes remains unclear. We enrolled 96 participants, including 46 with Mycobacterium avium complex (MAC)-LD, 23 with M. abscessus (MAB)-LD, and 27 controls. We measured expressions of PD-1, cytotoxic T-lymphocyte antigen-4 (CTLA-4) and regulatory T (Treg) cells on CD4+ lymphocytes and myeloid-derived suppressor cells (MDSCs) and analyzed their association with clinical features and radiographic outcomes. The percentage of PD-1 on CD4+(PD-1+CD4+) lymphocytes and MDSCs were higher in the MAC-LD group than the controls. There were no intergroup differences regarding CTLA-4+CD4+ lymphocytes. Higher PD-1+CD4+ lymphocytes were found in M. intracellulare- and M. avium-LD than in other MAC-LD. Positive sputum acid-fast stains and fibrocavitary radiographic lesions were correlated with elevated PD-1+CD4+ lymphocytes and Treg cells. The percentage of PD-1+CD4+ lymphocytes at the initial and 2 months of follow-up significantly predicted subsequent radiographic progression. As markers of immune tolerance, PD-1+CD4+ lymphocytes and MDSCs were higher in MAC-LD patients. The levels of PD-1+CD4+ and Treg cells were correlated with high mycobacteria bacilli burden in NTM-LD. Monitoring the expressions of PD-1+CD4+ lymphocytes may predict radiographic progression.