MilliporeSigma
  • Mutational analysis of cysteine residues of the insect odorant co-receptor (Orco) from Drosophila melanogaster reveals differential effects on agonist- and odorant-tuning receptor-dependent activation.
Mutational analysis of cysteine residues of the insect odorant co-receptor (Orco) from Drosophila melanogaster reveals differential effects on agonist- and odorant-tuning receptor-dependent activation.
The Journal of biological chemistry (2014-10-02)
Rebecca M Turner, Stephen L Derryberry, Brijesh N Kumar, Thomas Brittain, Laurence J Zwiebel, Richard D Newcomb, David L Christie
ABSTRACT
Insect odorant receptors are heteromeric odorant-gated cation channels comprising a conventional odorant-sensitive tuning receptor (ORx) and a highly conserved co-receptor known as Orco. Orco is found only in insects, and very little is known about its structure and the mechanism leading to channel activation. In the absence of an ORx, Orco forms homomeric channels that can be activated by a synthetic agonist, VUAA1. Drosophila melanogaster Orco (DmelOrco) contains eight cysteine amino acid residues, six of which are highly conserved. In this study, we replaced individual cysteine residues with serine or alanine and expressed Orco mutants in Flp-In 293 T-Rex cells. Changes in intracellular Ca(2+) levels were used to determine responses to VUAA1. Replacement of two cysteines (Cys-429 and Cys-449) in a predicted intracellular loop (ICL3), individually or together, gave variants that all showed similar increases in the rate of response and sensitivity to VUAA1 compared with wild-type DmelOrco. Kinetic modeling indicated that the response of the Orco mutants to VUAA1 was faster than wild-type Orco. The enhanced sensitivity and faster response of the Cys mutants was confirmed by whole-cell voltage clamp electrophysiology. In contrast to the results from direct agonist activation of Orco, the two cysteine replacement mutants when co-expressed with a tuning receptor (DmelOR22a) showed an ∼10-fold decrease in potency for activation by 2-methyl hexanoate. Our work has shown that intracellular loop 3 is important for Orco channel activation. Importantly, this study also suggests differences in the structural requirements for the activation of homomeric and heteromeric Orco channel complexes.
Sigma-Aldrich
Methyl hexanoate, 99%