MilliporeSigma
  • Suzuki-Miyaura cross-couplings of arenediazonium tetrafluoroborate salts with arylboronic acids catalyzed by aluminium hydroxide-supported palladium nanoparticles.
Suzuki-Miyaura cross-couplings of arenediazonium tetrafluoroborate salts with arylboronic acids catalyzed by aluminium hydroxide-supported palladium nanoparticles.
Organic & biomolecular chemistry (2011-11-22)
Xing Li, Xu-Ying Yan, Hong-Hong Chang, Li-Chao Wang, Yan Zhang, Wen-Wen Chen, Yan-Wei Li, Wen-Long Wei
ABSTRACT
Suzuki-Miyaura cross-couplings of arenediazonium salts with arylboronic acids catalyzed by highly active aluminium hydroxide-supported palladium nanoparticles catalyst have been investigated for the first time. The reactions are performed at 25 °C in MeOH without any base and ligand to afford biaryls in good to excellent yields under non-anhydrous and non-degassed conditions.
Sigma-Aldrich
4-Bromophenylboronic acid, ≥95.0%