MilliporeSigma
  • Targeting olfactomedin-like 3 inhibits tumor growth by impairing angiogenesis and pericyte coverage.
Targeting olfactomedin-like 3 inhibits tumor growth by impairing angiogenesis and pericyte coverage.
Molecular cancer therapeutics (2012-09-25)
Marijana Miljkovic-Licina, Philippe Hammel, Sarah Garrido-Urbani, Boris P-L Lee, Mehdi Meguenani, Chiraz Chaabane, Marie-Luce Bochaton-Piallat, Beat A Imhof
ABSTRACT
Antiangiogenic drugs have been used as anticancer agents to target tumor endothelial cells or pericytes. Because of limited efficacy of the current monotherapies, there is a strong demand for the dual targeting of endothelial cells and pericytes. Here, we identify Olfactomedin-like 3 (Olfml3) as a novel proangiogenic cue within the tumor microenvironment. Tumor-derived Olfml3 is produced by both tumor endothelial cells and accompanying pericytes and deposited in the perivascular compartment. Blockade of Olfml3 by anti-Olfml3 antibodies is highly effective in reducing tumor vascularization, pericyte coverage, and tumor growth. In vitro, Olfml3 targeting is sufficient to inhibit endothelioma cell migration and sprouting. Olfml3 alone or through binding to BMP4 enhances the canonical SMAD1/5/8 signaling pathway required for BMP4-induced angiogenesis. Therefore, Olfml3 blockade provides a novel strategy to control tumor growth by targeting two distinct cell types within the tumor microenvironment using a single molecule.
Sigma-Aldrich
Anti-NG2 Antibody, clone 132.38, clone 132.38, Upstate®, from mouse