MilliporeSigma
  • Human stem cells home to and repair laser-damaged trabecular meshwork in a mouse model.
Human stem cells home to and repair laser-damaged trabecular meshwork in a mouse model.
Communications biology (2018-12-12)
Hongmin Yun, Yiwen Wang, Yi Zhou, Ke Wang, Ming Sun, Donna B Stolz, Xiaobo Xia, C Ross Ethier, Yiqin Du
ABSTRACT
Glaucoma is the leading cause of irreversible vision loss, and reducing elevated intraocular pressure is currently the only effective clinical treatment. The trabecular meshwork is the main resistance site for aqueous outflow that maintains intraocular pressure. In this study, we transplanted human trabecular meshwork stem cells (TMSCs) intracamerally into mice that received laser photocoagulation over a 180° arc of the trabecular meshwork. TMSCs preferentially homed and integrated to the laser-damaged trabecular meshwork region and expressed differentiated cell markers at 2 and 4 weeks. Laser-induced inflammatory and fibrotic responses were prevented by TMSC transplantation with simultaneous ultrastructure and function restoration. Cell affinity and migration assays and elevated expression of CXCR4 and SDF1 in laser-treated mouse trabecular meshwork suggest that the CXCR4/SDF1 chemokine axis plays an important role in TMSC homing. Our results suggest that TMSCs may be a viable candidate for trabecular meshwork refunctionalization as a novel treatment for glaucoma.
Sigma-Aldrich
Anti-SDF-1 Antibody, clone K15C, Azide free, clone K15C, from mouse