MilliporeSigma
  • Exosomes in colorectal carcinoma formation: ALIX under the magnifying glass.
Exosomes in colorectal carcinoma formation: ALIX under the magnifying glass.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2016-05-07)
Gábor Valcz, Orsolya Galamb, Tibor Krenács, Sándor Spisák, Alexandra Kalmár, Árpád V Patai, Barna Wichmann, Kristóf Dede, Zsolt Tulassay, Béla Molnár
ABSTRACT
Exosomes are small membrane vesicles that have important roles in transporting a great variety of bioactive molecules between epithelial compartment and their microenvironment during tumor formation including colorectal adenoma-carcinoma sequence. We tested the mRNA expression of the top 25 exosome-related markers based on ExoCharta database in healthy (n=49), adenoma (n=49) and colorectal carcinoma (n=49) patients using Affymetrix HGU133 Plus2.0 microarrays. Most related genes showed significantly elevated expression including PGK1, PKM, ANXA5, ENO1, HSP90AB1 and MSN during adenoma-carcinoma sequence. Surprisingly, the expression of ALIX (ALG 2-interacting protein X), involved in multivesicular body (MVB) and exosome formation, was significantly reduced in normal vs adenoma (P=5.02 × 10(-13)) and in normal vs colorectal carcinoma comparisons (P=1.51 × 10(-10)). ALIX also showed significant reduction (P<0.05) at the in situ protein level in the epithelial compartment of adenoma (n=35) and colorectal carcinoma (n=37) patients compared with 27 healthy individuals. Furthermore, significantly reduced ALIX protein levels were accompanied by their gradual transition from diffuse cytoplasmic expression to granular signals, which fell into the 0.6-2 μm diameter size range of MVBs. These ALIX-positive particles were seen in the tumor nests, including tumor-stroma border, which suggest their exosome function. MVB-like structures were also detected in tumor microenvironment including α-smooth muscle actin-positive stromal cells, budding off cancer cells in the tumor front as well as in cancer cells entrapped within lymphoid vessels. In conclusion, we determined the top aberrantly expressed exosome-associated markers and revealed the transition of diffuse ALIX protein signals into a MVB-like pattern during adenoma-carcinoma sequence. These tumor-associated particles seen both in the carcinoma and the surrounding microenvironment can potentially mediate epithelial-stromal interactions involved in the regulation of tumor growth, metastatic invasion and therapy response.
Sigma-Aldrich
Anti-PDCD6IP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution