MilliporeSigma
  • Inhibition of Neutrophil Primary Granule Release during Yersinia pestis Pulmonary Infection.
Inhibition of Neutrophil Primary Granule Release during Yersinia pestis Pulmonary Infection.
mBio (2019-12-12)
Kara R Eichelberger, Grant S Jones, William E Goldman
ABSTRACT
Inhalation of Yersinia pestis causes primary pneumonic plague, the most severe manifestation of plague that is characterized by a dramatic neutrophil influx to the lungs. Neutrophils are ineffective during primary pneumonic plague, failing to control Y. pestis growth in the airways. However, the mechanisms by which Y. pestis resists neutrophil killing are incompletely understood. Here, we show that Y. pestis inhibits neutrophil degranulation, an important line of host innate immune defense. We observed that neutrophils from the lungs of mice infected intranasally with Y. pestis fail to release primary granules throughout the course of disease. Using a type III secretion system (T3SS) injection reporter strain, we determined that Y. pestis directly inhibits neutrophil granule release by a T3SS-dependent mechanism. Combinatorial mutant analysis revealed that a Y. pestis strain lacking both effectors YopE and YopH did not inhibit primary granule release and is killed by neutrophils both in vivo and in vitro Similarly, Y. pestis strains injecting only YopE or YopH are able to inhibit the majority of primary granule release from human neutrophils. We determined that YopE and YopH block Rac2 activation and calcium flux, respectively, to inhibit neutrophil primary granule release in isolated human neutrophils. These results demonstrate that Y. pestis coordinates the inhibition of neutrophil primary granule release through the activities of two distinct effectors, and this inhibition promotes Y. pestis survival during primary pneumonic plague.IMPORTANCEYersinia pestis is the causative agent of plague and is one of the deadliest human pathogens. The pneumonic form of Y. pestis infection has played a critical role in the severity of both historical and modern plague outbreaks, yet the host-pathogen interactions that govern the lethality of Yersinia pestis pulmonary infections are incompletely understood. Here, we report that Yersinia pestis inhibits neutrophil degranulation during infection, rendering neutrophils ineffective and allowing unrestricted growth of Y. pestis in the lungs. This coordinated inhibition of granule release not only demonstrates the pathogenic benefit of "silencing" lung neutrophils but also reveals specific host processes and pathways that could be manipulated to reduce the severity of primary pneumonic plague.
Sigma-Aldrich
EHT 1864, ≥98% (HPLC)