MilliporeSigma
  • Spinal cord injury and its underlying mechanism in rats with temporal lobe epilepsy.
Spinal cord injury and its underlying mechanism in rats with temporal lobe epilepsy.
Experimental and therapeutic medicine (2020-02-28)
Jinjie Liu, Zanhua Liu, Guoliang Liu, Kai Gao, Hengjie Zhou, Yongbo Zhao, Hong Wang, Lin Zhang, Sibo Liu
ABSTRACT
Numerous cases of spinal cord injury following seizure have been previously reported. However, whether spinal cord injury is a common occurrence after seizures and its underlying mechanisms remain unclear. The present study generated a Sprague-Dawley rat model of temporal lobe epilepsy (TLE), and Nissl staining and transmission electron microscopy were used to detect tissue damage. In addition, Evans blue staining was used to detect damage to the blood-brain barrier (BBB) and albumin extravasation. In addition, double-staining was used to detect the association between neurons and extravasated albumin. Furthermore, neuronal degeneration was assessed using Fluoro-Jade C staining, while fluorescence staining and western blotting were used to detect apoptosis and inflammation. In the present study, spinal cord injury was only observed in rats with grade IV-V seizures, whereas Nissl staining showed structural damage and decreased neuronal cell numbers in the brain and the spinal cord. The present study identified BBB damage and albumin extravasation in rats of the TLE groups. Double-staining for albumin and neurons showed a significant match of neurons positive for albumin. Fluoro-Jade C staining indicated neuronal degeneration in the brain, but not the spinal cord in the TLE rats. High levels of caspase-3 were also detected in the injured spinal cord. A small number of albumin+ neurons in the spinal cord presented caspase-3+ signals in rats of the TLE groups. The expression levels of intercellular adhesion molecule 1, CD11b and inflammatory factors such as tumor necrosis factor-α and interleukin-6 were significantly elevated in the injured spinal cord. The present results suggested that spinal cord injury occurred in rats as a result of severe seizure attacks, and that BBB damage, albumin extravasation, inflammation and apoptosis contributed to the pathological changes observed during spinal cord injury.
Sigma-Aldrich
Avidin–FITC from egg white, buffered aqueous solution